Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103511.1 | 4 | 752 | |||
XM_006713783.3 | XP_006713846.1 | 3 | 2156 | |||
XM_017007311.1 | XP_016862800.1 | 5 | 2335 | |||
XM_017007312.1 | XP_016862801.1 | 4 | 2390 | |||
XM_017007313.1 | XP_016862802.1 | 4 | 2391 | |||
GenBank mRNA | AK292958.1 | - | 3 | 416 | ||
AK310829.1 | - | 3 | 407 | |||
AL136815.1 | - | 2 | 183 | |||
BC024239.2 | - | 4 | 522 | 4826873 | ||
BG722744.1 | - | 4 | 527 | |||
BM705017.1 | - | 4 | 373 |