Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111901.1 | 9, 10 | 1556 | |||
GenBank mRNA | BC014983.1 | - | 9, 10 | 1506 | 4861869 |