Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319059.1 | NP_001305988.1 | 5 | 592 | ||
NM_145740.4 | NP_665683.1 | 6 | 725 | |||
XM_005249034.3 | XP_005249091.1 | 6 | 584 | |||
GenBank mRNA | AK293371.1 | - | 6 | 574 | ||
AK310324.1 | - | 6 | 725 | |||
AY532928.1 | - | 5 | 512 | |||
BC053578.1 | - | 6 | 560 | 5167338 | ||
BC110891.1 | - | 6 | 562 | 5171697 | ||
BI762344.1 | - | 6 | 601 | |||
CR407656.1 | - | 5 | 512 | |||
M14777.1 | - | 6 | 578 | |||
M15872.1 | - | 6 | 567 | |||
M21758.1 | - | 6 | 578 | |||
M25627.1 | - | 6 | 578 | |||
S49975.1 | - | 5 | 553 |