Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267551.1 | NP_001254480.1 | 2 | 119 | ||
NM_001267552.1 | NP_001254481.1 | 2 | 185 | |||
NM_007169.2 | NP_009100.2 | 2 | 177 | |||
NM_148172.2 | NP_680477.1 | 2 | 185 | |||
NM_148173.1 | NP_680478.1 | 2 | 64 | |||
XM_006721418.3 | XP_006721481.2 | 2 | 66 | |||
GenBank mRNA | AB029821.1 | - | 2 | 177 | ||
AF044214.1 | - | 2 | 150 | |||
AF113126.1 | - | 2 | 154 | |||
AF176806.1 | - | 2 | 127 | |||
AF176807.1 | - | 2 | 72 | |||
AK302251.1 | - | 2 | 190 | |||
BC000557.2 | - | 2 | 161 | 3162576 | ||
BC050593.1 | - | 2 | 162 | 6148983 | ||
BM011377.1 | - | 2 | 47 |