Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152486.2 | NP_689699.2 | 12, 13 | 1766 | ||
GenBank mRNA | AK054643.1 | - | 6, 7 | 1168 | ||
BC024295.2 | - | 8, 9 | 1341 | 3354502 | ||
BC033213.1 | - | 6, 7 | 1131 | 5014368 | ||
JX093066.1 | - | 11, 12 | 1641 | |||
JX093067.1 | - | 11, 12 | 1689 | |||
JX093068.1 | - | 11, 12 | 1689 | |||
JX093070.1 | - | 11, 12 | 1641 | |||
JX093071.1 | - | 11, 12 | 1638 | |||
JX093072.1 | - | 8, 9 | 1311 | |||
JX093073.1 | - | 11, 12 | 1689 | |||
JX093074.1 | - | 11, 12 | 1608 | |||
JX093076.1 | - | 10, 11 | 1500 | |||
JX093077.1 | - | 10, 11 | 1446 | |||
JX093078.1 | - | 10, 11 | 1880 | |||
JX093079.1 | - | 9, 10 | 1362 | |||
JX093080.1 | - | 10, 11 | 1835 | |||
JX093081.1 | - | 9, 10 | 1359 | |||
JX093082.1 | - | 10, 11 | 1832 | |||
JX093083.1 | - | 10, 11 | 1832 | |||
JX093084.1 | - | 8, 9 | 1556 | |||
JX093086.1 | - | 11, 12 | 1615 | |||
JX093087.1 | - | 11, 12 | 1555 | |||
JX093089.1 | - | 11, 12 | 1610 | |||
JX093090.1 | - | 11, 12 | 1604 | |||
JX093093.1 | - | 10, 11 | 1523 | |||
JX093094.1 | - | 10, 11 | 1478 | |||
JX093095.1 | - | 10, 11 | 1475 | |||
JX093096.1 | - | 9, 10 | 1291 | |||
JX093097.1 | - | 8, 9 | 1175 | |||
JX093098.1 | - | 5, 6 | 1200 | |||
JX093099.1 | - | 11, 12 | 1968 | |||
JX093100.1 | - | 12, 13 | 1771 | |||
JX093101.1 | - | 11, 12 | 1563 | |||
JX093102.1 | - | 5, 6 | 1076 | |||
JX093103.1 | - | 11, 12 | 1675 | |||
JX093104.1 | - | 8, 9 | 1777 | |||
JX093105.1 | - | 10, 11 | 1564 | |||
JX093106.1 | - | 9, 10 | 1350 | |||
JX093107.1 | - | 8, 9 | 1234 | |||
JX093108.1 | - | 10, 11 | 1513 | |||
JX093109.1 | - | 7, 8 | 1126 | |||
JX093110.1 | - | 8, 9 | 1293 |