Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098272.2 | NP_001091742.1 | 4 | 706 | ||
NM_001324219.1 | NP_001311148.1 | 3 | 693 | |||
NM_001324220.1 | NP_001311149.1 | 4 | 726 | |||
NM_002130.7 | NP_002121.4 | 3 | 647 | |||
XM_011514036.2 | XP_011512338.1 | 1 | 32 | |||
XM_017009412.1 | XP_016864901.1 | 4 | 786 | |||
GenBank mRNA | AK095492.1 | - | 4 | 514 | ||
AK315593.1 | - | 4 | 634 | |||
BC000297.2 | - | 4 | 585 | |||
BC083514.1 | - | 3 | 545 | 4443624 | ||
BT007302.1 | - | 2 | 480 | |||
L25798.1 | - | 2 | 501 | |||
X66435.1 | - | 4 | 602 |