Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083585.2 | NP_001077054.1 | 2 | 331 | ||
NM_001291582.1 | NP_001278511.1 | 2 | 331 | |||
NM_004703.5 | NP_004694.2 | 2 | 331 | |||
XM_011524060.1 | XP_011522362.1 | 2 | 331 | |||
XM_017025294.1 | XP_016880783.1 | 1 | 37 | |||
XM_017025295.1 | XP_016880784.1 | 1 | 37 | |||
GenBank mRNA | AF098638.1 | - | 2 | 107 | ||
AK314183.1 | - | 2 | 322 | |||
BC032435.1 | - | 2 | 306 | |||
BC041700.1 | - | 2 | 297 | 6046320 | ||
DB169027.1 | - | 2 | 331 | |||
X77723.1 | - | 2 | 180 | |||
X91141.1 | - | 2 | 295 |