Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281727.1 | NP_001268656.1 | 3 | 210 | ||
NM_001281728.1 | NP_001268657.1 | 4 | 289 | |||
NM_001281729.1 | NP_001268658.1 | 4 | 597 | |||
NM_001319157.1 | NP_001306086.1 | 3 | 214 | |||
NM_014183.3 | NP_054902.1 | 3 | 214 | |||
NM_177953.2 | NP_808852.1 | 4 | 293 | |||
NR_104032.1 | 4 | 296 | ||||
GenBank mRNA | AF132750.1 | - | 3 | 194 | ||
AF161511.1 | - | 3 | 185 | |||
AF165516.1 | - | 3 | 167 | |||
AF178431.1 | - | 2 | 139 | |||
AK026864.1 | - | 2 | 150 | |||
AK294214.1 | - | 3 | 172 | |||
AY026513.1 | - | 2 | 139 | |||
BC002481.1 | - | 3 | 192 | 3347555 | ||
BG110161.1 | - | 3 | 199 | |||
BG419592.1 | - | 4 | 265 | |||
BM561321.1 | - | 4 | 567 | |||
BM791453.1 | - | 4 | 275 | |||
BQ639804.1 | - | 4 | 262 | |||
BU163605.1 | - | 233 | ||||
CB149456.1 | - | 3 | 210 |