Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178026.2 | NP_821158.2 | 3 | 524 | ||
XM_017027809.1 | XP_016883298.1 | 3 | 523 | |||
XM_017027810.1 | XP_016883299.1 | 3 | 523 | |||
XM_017027811.1 | XP_016883300.1 | 3 | 523 | |||
GenBank mRNA | AK097100.1 | - | 2 | 382 | ||
AK124788.1 | - | 2 | 838 | |||
AK299494.1 | - | 3 | 522 | |||
AY138815.1 | - | 3 | 481 | |||
BC033745.1 | - | 3 | 524 | |||
BC040030.1 | - | 4 | 539 | |||
BC101371.1 | - | 3 | 513 | |||
BC101372.1 | - | 3 | 520 | |||
BC141843.1 | - | 3 | 513 | 8860314 | ||
BX401449.2 | - | 3 | 531 | |||
CA454346.1 | - | 3 | 520 |