Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002158.3 | NP_002149.2 | 3, 4 | 860 | ||
XM_005264282.2 | XP_005264339.1 | 2, 3 | 734 | |||
XM_005264283.2 | XP_005264340.1 | 4, 5 | 1055 | |||
XM_006712001.3 | XP_006712064.1 | 3, 4 | 791 | |||
XM_006712002.3 | XP_006712065.1 | 2, 3 | 751 | |||
XM_011532808.2 | XP_011531110.1 | 4, 5 | 977 | |||
XM_017003956.1 | XP_016859445.1 | 3, 4 | 1195 | |||
XM_017003957.1 | XP_016859446.1 | 4, 5 | 1052 | |||
XM_017003958.1 | XP_016859447.1 | 4, 5 | 1051 | |||
XM_017003959.1 | XP_016859448.1 | 4, 5 | 898 | |||
GenBank mRNA | BC063305.1 | - | 3, 4 | 785 | 30343577 | |
BC069731.1 | - | 1, 2 | 353 | |||
U57029.1 | - | 1, 2 | 556 |