Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFT2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145169.2 | NP_660152.1 | 3 | 329 | ||
NR_130112.1 | 4 | 352 | ||||
NR_130113.1 | 4 | 352 | ||||
NR_130114.1 | 4 | 343 | ||||
GenBank mRNA | AF041429.1 | - | 3 | 263 | ||
AW964442.1 | - | 4 | 274 | |||
BC018969.1 | - | 3 | 234 | 3959886 | ||
BC028388.1 | - | 4 | 296 | |||
BF223532.1 | - | 4 | 247 | |||
BG706927.1 | - | 3 | 318 | |||
BI597156.1 | - | 4 | 314 | |||
DB476616.1 | - | 4 | 352 |