Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004195.2 | NP_004186.1 | 5 | 1065 | ||
NM_148901.1 | NP_683699.1 | 4 | 854 | |||
NM_148902.1 | NP_683700.1 | 5 | 1044 | |||
XM_017002722.1 | XP_016858211.1 | 4 | 2321 | |||
GenBank mRNA | AF241229.1 | - | 4 | 716 | ||
KP067911.1 | - | 4 | 1130 |