Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152486.2 | NP_689699.2 | 14 | 1910 | ||
GenBank mRNA | AK054643.1 | - | 7 | 1411 | ||
BC024295.2 | - | 10 | 1485 | 3354502 | ||
BC033213.1 | - | 8 | 1275 | 5014368 | ||
JX093066.1 | - | 12 | 1884 | |||
JX093067.1 | - | 13 | 1848 | |||
JX093068.1 | - | 13 | 1833 | |||
JX093069.1 | - | 12 | 1657 | |||
JX093070.1 | - | 13 | 1785 | |||
JX093071.1 | - | 13 | 1782 | |||
JX093072.1 | - | 10 | 1455 | |||
JX093074.1 | - | 13 | 1752 | |||
JX093076.1 | - | 12 | 1644 | |||
JX093077.1 | - | 12 | 1590 | |||
JX093078.1 | - | 12 | 2024 | |||
JX093079.1 | - | 11 | 1506 | |||
JX093080.1 | - | 11 | 2078 | |||
JX093081.1 | - | 11 | 1503 | |||
JX093082.1 | - | 12 | 1976 | |||
JX093083.1 | - | 11 | 2075 | |||
JX093084.1 | - | 10 | 1700 | |||
JX093085.1 | - | 8 | 802 | |||
JX093086.1 | - | 13 | 1759 | |||
JX093087.1 | - | 13 | 1699 | |||
JX093088.1 | - | 8 | 799 | |||
JX093089.1 | - | 13 | 1754 | |||
JX093090.1 | - | 13 | 1748 | |||
JX093091.1 | - | 5 | 725 | |||
JX093094.1 | - | 12 | 1622 | |||
JX093095.1 | - | 12 | 1619 | |||
JX093096.1 | - | 11 | 1435 | |||
JX093097.1 | - | 10 | 1319 | |||
JX093098.1 | - | 7 | 1344 | |||
JX093099.1 | - | 13 | 2112 | |||
JX093100.1 | - | 14 | 1915 | |||
JX093101.1 | - | 13 | 1707 | |||
JX093102.1 | - | 7 | 1220 | |||
JX093103.1 | - | 13 | 1819 | |||
JX093104.1 | - | 10 | 1921 | |||
JX093105.1 | - | 12 | 1708 | |||
JX093106.1 | - | 11 | 1494 | |||
JX093107.1 | - | 10 | 1378 | |||
JX093108.1 | - | 12 | 1657 | |||
JX093109.1 | - | 9 | 1270 | |||
JX093110.1 | - | 10 | 1437 |