Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003782.3 | NP_003773.1 | 1 | 1668 | ||
GenBank mRNA | AB026730.1 | - | 1 | 1638 | ||
AF063595.1 | - | 1 | 497 | |||
BC032574.1 | - | 1 | 1649 | 5247350 |