Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281727.1 | NP_001268656.1 | 3, 4 | 326 | ||
NM_001281728.1 | NP_001268657.1 | 4, 5 | 405 | |||
NM_001281729.1 | NP_001268658.1 | 4, 5 | 713 | |||
NM_014183.3 | NP_054902.1 | 3, 4 | 330 | |||
NM_177953.2 | NP_808852.1 | 4, 5 | 409 | |||
NR_104032.1 | 4, 5 | 412 | ||||
GenBank mRNA | AF132750.1 | - | 3, 4 | 310 | ||
AF161511.1 | - | 3, 4 | 301 | |||
AF165516.1 | - | 3, 4 | 283 | |||
AF178431.1 | - | 2, 3 | 255 | |||
AK026864.1 | - | 2, 3 | 266 | |||
AY026513.1 | - | 2, 3 | 255 | |||
BC002481.1 | - | 3, 4 | 308 | 3347555 | ||
BG110161.1 | - | 3, 4 | 315 | |||
BG419592.1 | - | 4, 5 | 381 | |||
BM561321.1 | - | 4, 5 | 683 | |||
BM791453.1 | - | 4, 5 | 391 | |||
BQ639804.1 | - | 4, 5 | 378 | |||
BU163605.1 | - | 349 | ||||
CB149456.1 | - | 3, 4 | 326 |