Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144628.3 | NP_653229.1 | 8 | 4192 | ||
NR_111901.1 | 10 | 1772 | ||||
XM_005260661.1 | XP_005260718.1 | 9 | 3231 | |||
XM_006723540.3 | XP_006723603.1 | 7 | 3971 | |||
XM_017027645.1 | XP_016883134.1 | 8 | 3012 | |||
GenBank mRNA | AF116909.1 | - | 1 | 622 | ||
AK092063.1 | - | 1 | 1978 | |||
BC014983.1 | - | 10 | 1722 | 4861869 | ||
BX647370.1 | - | 3 | 4923 |