Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042518.1 | NP_001035983.1 | 2 | 245 | ||
NM_175739.3 | NP_783866.2 | 2 | 245 | |||
XM_011536714.2 | XP_011535016.1 | 2 | 351 | |||
XM_011536715.2 | XP_011535017.1 | 3 | 215 | |||
GenBank mRNA | AK301082.1 | - | 2 | 139 | ||
AY185496.1 | - | 2 | 245 | |||
AY220118.1 | - | 2 | 159 | |||
AY220119.1 | - | 2 | 123 | |||
AY358700.1 | - | 2 | 89 | |||
BC111498.1 | - | 2 | 148 | 5498091 |