Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHSY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175856.4 | NP_787052.3 | 2 | 1011 | ||
XM_005271982.3 | XP_005272039.1 | 2 | 1385 | |||
XM_005271983.3 | XP_005272040.1 | 2 | 1385 | |||
XM_011543363.2 | XP_011541665.1 | 2 | 864 | |||
XM_011543365.2 | XP_011541667.1 | 2 | 1385 | |||
XM_017009435.1 | XP_016864924.1 | 2 | 1385 | |||
GenBank mRNA | AB175496.1 | - | 2 | 1011 | ||
AJ504664.2 | - | 2 | 548 | |||
AJ578034.2 | - | 2 | 1369 | |||
AJ862728.1 | - | 2 | 1009 | |||
AY726604.1 | - | 2 | 313 | |||
BC137325.1 | - | 2 | 1080 | 9021322 |