Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021630.5 | NP_067643.3 | 3 | 936 | ||
NM_176871.3 | NP_789847.1 | 2 | 278 | |||
NM_198042.3 | NP_932159.1 | 2 | 278 | |||
GenBank mRNA | AF289563.1 | - | 3 | 535 | ||
AK024498.1 | - | 2 | 221 | |||
AK027800.1 | - | 3 | 547 | |||
AK092968.1 | - | 3 | 936 | |||
AY007729.1 | - | 2 | 302 | |||
AY070438.1 | - | 2 | 302 | |||
AY217349.1 | - | 2 | 263 | |||
BC021556.1 | - | 3 | 417 | 3959372 | ||
BC071774.1 | - | 3 | 547 | 4593207 | ||
DN997519.1 | - | 3 | 222 |