Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173039.2 | NP_766627.1 | 1 | 834 | ||
XM_005273917.4 | XP_005273974.1 | 1 | 815 | |||
GenBank mRNA | AB028147.1 | - | 1 | 781 | ||
AK075346.1 | - | 1 | 476 | |||
BC040443.1 | - | 1 | 834 | 5728157 |