Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193421.1 | NP_001180350.1 | 2 | 469 | ||
NM_173485.5 | NP_775756.3 | 2 | 1002 | |||
XM_017027640.1 | XP_016883129.1 | 2 | 1100 | |||
XM_017027641.1 | XP_016883130.1 | 2 | 1100 | |||
GenBank mRNA | AK091206.1 | - | 2 | 195 | ||
AK302570.1 | - | 2 | 469 | |||
BX640770.1 | - | 2 | 1002 |