Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175735.3 | NP_783862.2 | 4 | 334 | ||
XM_017003751.1 | XP_016859240.1 | 5 | 1499 | |||
GenBank mRNA | AF323919.1 | - | 4 | 359 | ||
AY026238.1 | - | 4 | 359 | |||
BC100882.3 | - | 3 | 242 | 40003264 | ||
BC100883.2 | - | 3 | 242 | 40003266 | ||
BC100884.1 | - | 3 | 242 | 40003273 | ||
BC100885.2 | - | 3 | 242 | 40003274 | ||
BQ187749.1 | - | 5 | 388 |