Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHSY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175856.4 | NP_787052.3 | 3 | 1136 | ||
XM_011543363.2 | XP_011541665.1 | 3 | 989 | |||
XM_011543364.2 | XP_011541666.1 | 2 | 345 | |||
XM_011543365.2 | XP_011541667.1 | 4 | 1598 | |||
XM_017009434.1 | XP_016864923.1 | 2 | 10035 | |||
GenBank mRNA | AB086062.1 | - | 3 | 1136 | ||
AB175496.1 | - | 3 | 1136 | |||
AJ504664.2 | - | 3 | 673 | |||
AJ578034.2 | - | 3 | 1494 | |||
AY726604.1 | - | 3 | 438 | |||
BC137325.1 | - | 3 | 1205 | 9021322 |