Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKHD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177531.4 | NP_803875.2 | 69 | 11222 | ||
XM_011517371.2 | XP_011515673.1 | 69 | 11173 | |||
XM_017013969.1 | XP_016869458.1 | 69 | 11173 | |||
XM_017013970.1 | XP_016869459.1 | 68 | 11059 | |||
XM_017013971.1 | XP_016869460.1 | 69 | 11011 | |||
XM_017013972.1 | XP_016869461.1 | 67 | 10870 | |||
XM_017013973.1 | XP_016869462.1 | 45 | 8229 | |||
XM_017013974.1 | XP_016869463.1 | 40 | 7729 | |||
GenBank mRNA | AK131338.1 | - | 15 | 1904 | ||
AY219181.1 | - | 69 | 11222 |