Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003712.3 | NP_003703.1 | 3 | 447 | ||
NM_177526.2 | NP_803545.1 | 3 | 406 | |||
NM_177543.2 | NP_808211.1 | 3 | 514 | |||
XM_011528396.2 | XP_011526698.1 | 3 | 443 | |||
GenBank mRNA | AB209696.1 | - | 1 | 933 | ||
AF035959.1 | - | 3 | 391 | |||
AF047760.1 | - | 3 | 391 | |||
AF056083.1 | - | 3 | 391 | |||
BC002806.2 | - | 3 | 398 | 3659728 | ||
BE615792.1 | - | 3 | 468 | |||
BG831889.1 | - | 3 | 515 | |||
BM050291.1 | - | 3 | 448 | |||
BT007021.1 | - | 3 | 348 | |||
HY160303.1 | - | 3 | 373 |