Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001315501.1 | NP_001302430.1 | 7, 8 | 1202 | ||
NM_001315502.1 | NP_001302431.1 | 5, 6 | 692 | |||
NM_002555.5 | NP_002546.3 | 7, 8 | 986 | |||
NM_183233.2 | NP_899056.2 | 7, 8 | 989 | |||
XM_011520141.2 | XP_011518443.2 | 7, 8 | 1145 | |||
XM_011520142.2 | XP_011518444.2 | 7, 8 | 1144 | |||
GenBank mRNA | AB012083.1 | - | 7, 8 | 965 | ||
AF028738.1 | - | 7, 8 | 965 | |||
AF030302.1 | - | 7, 8 | 1202 | |||
AF037064.1 | - | 7, 8 | 973 | |||
AF059663.1 | - | 7, 8 | 965 | |||
AF070479.1 | - | 7, 8 | 965 | |||
AL521035.3 | - | 5, 6 | 689 | |||
AL832892.1 | - | 2, 3 | 711 | |||
BC015571.2 | - | 7, 8 | 989 | 3636077 |