Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002558.3 | NP_002549.1 | 6 | 992 | ||
XM_011523896.2 | XP_011522198.1 | 7 | 1384 | |||
XM_011523897.2 | XP_011522199.1 | 6 | 832 | |||
XM_011523899.2 | XP_011522201.1 | 6 | 1496 | |||
XM_011523900.2 | XP_011522202.1 | 6 | 1668 | |||
GenBank mRNA | AF020498.1 | - | 6 | 827 | ||
AK313264.1 | - | 6 | 791 | |||
BC027949.1 | - | 5 | 1074 | |||
BC044657.1 | - | 6 | 767 | 5756349 | ||
U45448.1 | - | 6 | 747 | |||
X83688.1 | - | 6 | 724 |