Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321931.1 | NP_001308860.1 | 6 | 1120 | ||
NM_001321932.1 | NP_001308861.1 | 6 | 1206 | |||
NM_001321933.1 | NP_001308862.1 | 6 | 1116 | |||
NM_001321934.1 | NP_001308863.1 | 5 | 879 | |||
NM_001321935.1 | NP_001308864.1 | 5 | 732 | |||
NM_001321936.1 | NP_001308865.1 | 6 | 1059 | |||
NM_001321938.1 | NP_001308867.1 | 7 | 1107 | |||
NM_001321939.1 | NP_001308868.1 | 4 | 1114 | |||
NM_001321940.1 | NP_001308869.1 | 6 | 1462 | |||
NM_001321941.1 | NP_001308870.1 | 6 | 1294 | |||
NM_001321942.1 | NP_001308871.1 | 6 | 1301 | |||
NM_001321943.1 | NP_001308872.1 | 6 | 1338 | |||
NM_001321944.1 | NP_001308873.1 | 6 | 1554 | |||
NM_001321945.1 | NP_001308874.1 | 6 | 1097 | |||
NM_001321946.1 | NP_001308875.1 | 7 | 1203 | |||
NM_001321947.1 | NP_001308876.1 | 6 | 1021 | |||
NM_001321948.1 | NP_001308877.1 | 6 | 1093 | |||
NM_001321949.1 | NP_001308878.1 | 6 | 1790 | |||
NM_004115.3 | NP_004106.1 | 5 | 856 | |||
NM_175929.2 | NP_787125.1 | 5 | 967 | |||
GenBank mRNA | AY188178.1 | - | 5 | 967 |