Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297696.1 | NP_001284625.1 | 3, 4 | 440 | ||
NM_001297697.1 | NP_001284626.1 | 4, 5 | 321 | |||
NM_176816.4 | NP_789786.2 | 3, 4 | 482 | |||
XM_005248458.4 | XP_005248515.1 | 3, 4 | 719 | |||
XM_005248459.4 | XP_005248516.1 | 3, 4 | 521 | |||
XM_005248464.3 | XP_005248521.1 | 3, 4 | 524 | |||
XM_006714569.3 | XP_006714632.1 | 3, 4 | 524 | |||
XM_006714570.3 | XP_006714633.1 | 3, 4 | 293 | |||
XM_011543255.2 | XP_011541557.1 | 3, 4 | 499 | |||
XM_011543256.2 | XP_011541558.1 | 3, 4 | 500 | |||
XM_011543257.2 | XP_011541559.1 | 3, 4 | 524 | |||
XM_011543258.2 | XP_011541560.1 | 3, 4 | 524 | |||
XM_011543259.2 | XP_011541561.1 | 3, 4 | 524 | |||
XM_011543260.2 | XP_011541562.1 | 3, 4 | 524 | |||
XM_017009206.1 | XP_016864695.1 | 3, 4 | 714 | |||
XM_017009207.1 | XP_016864696.1 | 3, 4 | 524 | |||
XM_017009209.1 | XP_016864698.1 | 3, 4 | 521 | |||
GenBank mRNA | AB024691.1 | - | 3, 4 | 482 | ||
AB024692.1 | - | 2, 3 | 185 | |||
AK299871.1 | - | 4, 5 | 321 | |||
BC108715.1 | - | 2, 3 | 416 | |||
BQ923745.1 | - | 2, 3 | 414 |