Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136504.1 | NP_001129976.1 | 5 | 768 | ||
NM_177402.4 | NP_796376.2 | 5 | 758 | |||
XM_011509192.2 | XP_011507494.1 | 5 | 788 | |||
XM_017000309.1 | XP_016855798.1 | 4 | 2534 | |||
XM_017000310.1 | XP_016855799.1 | 4 | 2525 | |||
XM_017000311.1 | XP_016855800.1 | 5 | 777 | |||
XM_017000312.1 | XP_016855801.1 | 5 | 1442 | |||
XM_017000313.1 | XP_016855802.1 | 5 | 1432 | |||
GenBank mRNA | AK090672.1 | - | 5 | 752 | ||
AK094430.1 | - | 5 | 768 | |||
BC100814.1 | - | 5 | 623 | 40000996 | ||
BC100815.1 | - | 5 | 623 | 40000997 | ||
BC100816.1 | - | 3 | 270 | |||
BC100817.1 | - | 5 | 623 | 40001000 |