Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKHD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177531.4 | NP_803875.2 | 73 | 11866 | ||
XM_011517371.2 | XP_011515673.1 | 73 | 11817 | |||
XM_017013969.1 | XP_016869458.1 | 73 | 11817 | |||
XM_017013970.1 | XP_016869459.1 | 72 | 11703 | |||
XM_017013971.1 | XP_016869460.1 | 73 | 11655 | |||
XM_017013972.1 | XP_016869461.1 | 71 | 11514 | |||
XM_017013973.1 | XP_016869462.1 | 49 | 8873 | |||
XM_017013974.1 | XP_016869463.1 | 44 | 8373 | |||
GenBank mRNA | AK131338.1 | - | 19 | 2548 | ||
AL133640.1 | - | 3 | 306 | |||
AY219181.1 | - | 73 | 11866 |