Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123369.1 | NP_001116841.1 | 1, 2 | 304 | ||
NM_002721.4 | NP_002712.1 | 1, 2 | 304 | |||
GenBank mRNA | AF035158.2 | - | 1, 2 | 83 | ||
AK295752.1 | - | 1, 2 | 124 | |||
AK299087.1 | - | 1, 2 | 168 | |||
AK312332.1 | - | 1, 2 | 136 | |||
AK316527.1 | - | 1, 2 | 158 | |||
BC006990.1 | - | 1, 2 | 151 | 3684339 | ||
BT019708.1 | - | 1, 2 | 83 | |||
DB037210.1 | - | 1, 2 | 304 | |||
DC369815.1 | - | 1, 2 | 168 | |||
KU178271.1 | - | 1, 2 | 83 | |||
KU178272.1 | - | 1, 2 | 83 | |||
X92972.2 | - | 1, 2 | 104 |