Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEA15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297576.1 | NP_001284505.1 | 4 | 619 | ||
NM_001297577.1 | NP_001284506.1 | 3 | 417 | |||
NM_003768.4 | NP_003759.1 | 2 | 312 | |||
XM_006711599.2 | XP_006711662.1 | 2 | 216 | |||
GenBank mRNA | AK095879.1 | - | 4 | 524 | ||
AK311762.1 | - | 2 | 241 | |||
BC002426.2 | - | 2 | 188 | 3346270 | ||
BC022554.1 | - | 2 | 299 | 4820225 | ||
BT007252.1 | - | 1 | 106 | |||
CR541733.1 | - | 1 | 106 | |||
DA131259.1 | - | 2 | 296 | |||
DA532423.1 | - | 2 | 296 | |||
DA707281.1 | - | 3 | 344 | |||
HY182028.1 | - | 2 | 315 | |||
X86809.1 | - | 2 | 219 | |||
Y13736.1 | - | 2 | 175 |