Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004637.5 | NP_004628.4 | 4 | 618 | ||
GenBank mRNA | AF498942.1 | - | 3 | 386 | ||
AK225177.1 | - | 3 | 372 | |||
AK225691.1 | - | 3 | 365 | |||
AK290721.1 | - | 4 | 522 | |||
AU122058.1 | - | 4 | 617 | |||
BC008721.2 | - | 4 | 561 | 2821435 | ||
BC013728.1 | - | 3 | 401 | 3882701 | ||
BX537775.1 | - | 2 | 334 |