Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005105.4 | NP_005096.1 | 6 | 609 | ||
GenBank mRNA | AF127761.1 | - | 6 | 511 | ||
AF161463.1 | - | 6 | 523 | |||
AF182415.1 | - | 6 | 528 | |||
AF198620.1 | - | 6 | 530 | |||
AF231511.1 | - | 6 | 516 | |||
AF299118.1 | - | 6 | 499 | |||
AK075009.1 | - | 6 | 530 | |||
BC017088.2 | - | 6 | 571 | 3850355 | ||
BC071577.1 | - | 6 | 557 | |||
CR541805.1 | - | 6 | 499 | |||
CR541823.1 | - | 6 | 499 | |||
KF051020.1 | - | 6 | 499 |