Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005517.3 | NP_005508.1 | 5, 6 | 447 | ||
GenBank mRNA | AK026562.1 | - | 5, 6 | 289 | ||
BC000378.1 | - | 5, 6 | 350 | 2820349 | ||
BC003050.1 | - | 5, 6 | 340 | 3543809 | ||
BC003689.1 | - | 5, 6 | 311 | |||
BC007124.1 | - | 1 | 137 | |||
BC014644.1 | - | 5, 6 | 330 | 3355317 | ||
BC032140.2 | - | 5, 6 | 339 | 5021546 | ||
BC061898.1 | - | 1 | 75 | |||
BC064347.1 | - | 1 | 117 | |||
BC070297.1 | - | 2 | 298 | 4282630 | ||
BC071707.1 | - | 5, 6 | 340 | 3849426 | ||
BC072010.1 | - | 5, 6 | 334 | 6273507 | ||
BC072011.1 | - | 5, 6 | 330 | 6647282 | ||
BC075837.1 | - | 5, 6 | 435 | 6272425 | ||
BC081567.1 | - | 5, 6 | 333 | 4131303 | ||
BC110390.1 | - | 5, 6 | 343 | 5190365 | ||
CR542122.1 | - | 1 | 245 | |||
M12623.1 | - | 5, 6 | 333 |
Gene Symbol: RPL12P38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090983.1 | - | 1 | 802 |