Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317893.1 | NP_001304822.1 | 2 | 285 | ||
NM_001317894.1 | NP_001304823.1 | 2 | 285 | |||
NM_014506.2 | NP_055321.1 | 2 | 285 | |||
NR_133948.1 | 2 | 285 | ||||
NR_134113.1 | 2 | 285 | ||||
XM_005251927.3 | XP_005251984.1 | 2 | 280 | |||
GenBank mRNA | AF317129.1 | - | 2 | 275 | ||
BC015578.2 | - | 2 | 230 | 4589479 | ||
BP366330.1 | - | 2 | 285 |