Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016134.3 | NP_057218.1 | 2 | 282 | ||
XM_017012953.1 | XP_016868442.1 | 2 | 282 | |||
XM_017012954.1 | XP_016868443.1 | 2 | 282 | |||
XM_017012955.1 | XP_016868444.1 | 2 | 282 | |||
GenBank mRNA | AF107834.1 | - | 1 | 125 | ||
AF119386.1 | - | 1 | 96 | |||
AK075132.1 | - | 2 | 200 | |||
AK296013.1 | - | 2 | 145 | |||
AK304008.1 | - | 2, 3 | 199 | |||
AK315447.1 | - | 2 | 199 | |||
BC020689.1 | - | 2 | 238 | 4251802 | ||
DA699663.1 | - | 2 | 282 |