Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291997.1 | NP_001278926.1 | 1 | 458 | ||
NM_016373.3 | NP_057457.1 | 1 | 458 | |||
NM_130791.3 | NP_570607.1 | 1 | 458 | |||
NR_120435.1 | 1 | 458 | ||||
NR_120436.1 | 1 | 458 | ||||
XM_011523101.2 | XP_011521403.1 | 1 | 217 | |||
XM_011523103.2 | XP_011521405.1 | 1 | 217 | |||
XM_011523104.2 | XP_011521406.1 | 1 | 217 | |||
XM_011523105.2 | XP_011521407.1 | 1 | 217 | |||
XM_017023278.1 | XP_016878767.1 | 1 | 217 | |||
GenBank mRNA | AF187015.1 | - | 1 | 176 | ||
AF211943.1 | - | 1 | 217 | |||
AF227526.1 | - | 1 | 177 | |||
AF227527.1 | - | 1 | 196 | |||
AF227528.1 | - | 1 | 193 | |||
AF227529.1 | - | 1 | 196 | |||
AF395123.1 | - | 1 | 225 | |||
AF395124.1 | - | 1 | 225 | |||
AK027626.1 | - | 1 | 216 | |||
AK290438.1 | - | 1 | 208 | |||
AK291300.1 | - | 1 | 215 | |||
AK298322.1 | - | 1 | 245 | |||
AW874692.1 | - | 1 | 103 | |||
AW874693.1 | - | 1 | 437 | |||
AY256821.1 | - | 1 | 147 | |||
BC003184.1 | - | 1 | 171 | 3535809 | ||
BT007445.1 | - | 92 | ||||
BU157392.1 | - | 1 | 85 | |||
DB035405.1 | - | 1 | 458 |