Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIKESHI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322404.1 | NP_001309333.1 | 2 | 325 | ||
NM_001322407.1 | NP_001309336.1 | 3 | 499 | |||
NM_001322409.1 | NP_001309338.1 | 3 | 429 | |||
NM_016401.3 | NP_057485.2 | 2 | 341 | |||
NR_136324.1 | 2 | 325 | ||||
XM_017017914.1 | XP_016873403.1 | 2 | 331 | |||
XM_017017915.1 | XP_016873404.1 | 2 | 118 | |||
GenBank mRNA | AF161487.1 | - | 2 | 325 | ||
AF161527.1 | - | 2 | 324 | |||
AK001447.1 | - | 2 | 312 | |||
AK223198.1 | - | 2 | 312 | |||
AK298763.1 | - | 2 | 301 | |||
BC001677.2 | - | 2 | 214 | 2820772 | ||
BC006476.2 | - | 2 | 214 | 2820772 | ||
BC015991.2 | - | 2 | 306 | 2820691 | ||
BC018080.1 | - | 2 | 275 | 4798958 | ||
BC021621.1 | - | 2 | 306 | 2820691 | ||
BX332668.2 | - | 2 | 178 |