Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040113.1 | NP_001035202.1 | 2 | 261 | ||
NM_001040114.1 | NP_001035203.1 | 2 | 261 | |||
NM_002474.2 | NP_002465.1 | 2 | 261 | |||
NM_022844.2 | NP_074035.1 | 2 | 261 | |||
XM_011522502.2 | XP_011520804.1 | 2 | 263 | |||
XM_017023250.1 | XP_016878739.1 | 2 | 225 | |||
GenBank mRNA | AB020673.2 | - | 2 | 229 | ||
AK308389.1 | - | 2 | 258 | |||
AY520816.1 | - | 2 | 242 | |||
AY520817.1 | - | 2 | 242 | |||
BC031040.1 | - | 2 | 259 | |||
BC101677.1 | - | 2 | 225 | 8069183 | ||
BC104906.1 | - | 2 | 225 | 8143909 | ||
BC143364.1 | - | 2 | 225 | 9051869 | ||
BX489788.1 | - | 2 | 261 | |||
EU489063.1 | - | 2 | 261 |