Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005880.3 | NP_005871.1 | 3 | 316 | ||
GenBank mRNA | AJ001309.1 | - | 3 | 226 | ||
AK313031.1 | - | 3 | 264 | |||
BC013044.2 | - | 3 | 268 | 3505399 | ||
BC015809.2 | - | 3 | 269 | 3922477 | ||
DB097731.1 | - | 3 | 390 |