Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: API5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142930.1 | NP_001136402.1 | 2 | 257 | ||
NM_001243747.1 | NP_001230676.1 | 2 | 257 | |||
NM_006595.3 | NP_006586.1 | 2 | 257 | |||
NR_024625.1 | 3 | 319 | ||||
XM_006718359.3 | XP_006718422.1 | 2 | 247 | |||
XM_017018464.1 | XP_016873953.1 | 3 | 304 | |||
XM_017018465.1 | XP_016873954.1 | 3 | 304 | |||
GenBank mRNA | AF229254.1 | - | 2 | 197 | ||
AK293303.1 | - | 2 | 158 | |||
AK299288.1 | - | 2 | 223 | |||
AK304157.1 | - | 3 | 285 | |||
AY311389.1 | - | 2 | 196 | |||
BC017709.1 | - | 2 | 177 | 4419401 | ||
BM193468.1 | - | 2 | 239 | |||
BT007093.1 | - | 2 | 84 | |||
DA104687.1 | - | 2 | 258 | |||
DC334531.1 | - | 2 | 257 | |||
U83857.1 | - | 2 | 179 | |||
Y15906.1 | - | 2 | 216 |