Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007179.2 | NP_009110.2 | 1 | 180 | ||
XM_011517702.2 | XP_011516004.1 | 1 | 268 | |||
GenBank mRNA | AF135824.1 | - | 1 | 164 | ||
AF156094.1 | - | 1 | 180 | |||
BC126473.1 | - | 1 | 189 | 8992189 | ||
BC126475.1 | - | 1 | 189 | 8992191 |