Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001279353.1 | NP_001266282.1 | 2 | 374 | ||
NM_001279354.1 | NP_001266283.1 | 2 | 363 | |||
NM_007259.4 | NP_009190.2 | 2 | 718 | |||
NR_103998.1 | 2 | 343 | ||||
XM_017000164.1 | XP_016855653.1 | 2 | 268 | |||
GenBank mRNA | AF165513.1 | - | 2 | 198 | ||
AJ133421.1 | - | 2 | 186 | |||
AK023170.1 | - | 2 | 205 | |||
AK223214.1 | - | 2 | 171 | |||
AK293555.1 | - | 2 | 204 | |||
AK295529.1 | - | 2 | 218 | |||
AK298786.1 | - | 2 | 285 | |||
AK301994.1 | - | 2 | 325 | |||
AK307949.1 | - | 2 | 164 | |||
BC012932.1 | - | 2 | 204 | 3853152 | ||
BC028382.2 | - | 2 | 699 | 4818323 | ||
BG176899.1 | - | 2 | 374 | |||
DA374440.1 | - | 2 | 343 | |||
U35246.1 | - | 2 | 196 |