Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297437.1 | NP_001284366.1 | 2 | 309 | ||
NM_012176.2 | NP_036308.1 | 2 | 307 | |||
NM_033484.2 | NP_277019.1 | 2 | 307 | |||
XM_011514026.2 | XP_011512328.1 | 2 | 312 | |||
XM_011514027.2 | XP_011512329.1 | 2 | 239 | |||
GenBank mRNA | AF129534.1 | - | 2 | 251 | ||
AF176703.1 | - | 2 | 245 | |||
AK001003.1 | - | 1 | 1785 | |||
AK024075.1 | - | 2 | 307 | |||
AU143782.1 | - | 2 | 307 | |||
BC040011.1 | - | 2 | 268 | |||
BC048098.1 | - | 2 | 275 | 4830192 |