Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013336.3 | NP_037468.1 | 2, 3 | 261 | ||
GenBank mRNA | AF077032.1 | - | 2, 3 | 167 | ||
AF084458.1 | - | 2, 3 | 167 | |||
AF346602.1 | - | 2, 3 | 143 | |||
AK027423.1 | - | 2, 3 | 261 | |||
AK074706.1 | - | 2, 3 | 261 | |||
AK299118.1 | - | 2, 3 | 341 | |||
AL832821.1 | - | 2, 3 | 194 | |||
BC002951.1 | - | 2, 3 | 168 |