Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002715.2 | NP_002706.1 | 2 | 547 | ||
GenBank mRNA | AB451240.1 | - | 2 | 149 | ||
AB451365.1 | - | 2 | 149 | |||
AK097599.1 | - | 2 | 359 | |||
BC000400.2 | - | 2 | 348 | 2822635 | ||
BC002657.2 | - | 2 | 348 | 3607862 | ||
BC019275.1 | - | 2 | 348 | 2822635 | ||
BC031696.1 | - | 2 | 348 | 5001649 | ||
CR457417.1 | - | 2 | 149 | |||
J03804.1 | - | 2 | 331 | |||
X12646.1 | - | 2 | 232 |
Gene Symbol: CDKL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK294559.1 | - | 10 | 1273 | ||
AK297586.1 | - | 7 | 951 |