Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300860.1 | NP_001287789.1 | 2 | 298 | ||
NM_014829.3 | NP_055644.2 | 2 | 298 | |||
NR_125341.1 | 2 | 298 | ||||
XM_011543758.1 | XP_011542060.1 | 2 | 261 | |||
GenBank mRNA | AB018344.1 | - | 2 | 237 | ||
AF106680.1 | - | 2 | 241 | |||
AK126011.1 | - | 2 | 253 | |||
AK291788.1 | - | 2 | 251 | |||
BC012304.2 | - | 2 | 228 | 3873814 | ||
BC105598.1 | - | 2 | 255 | |||
BF244218.1 | - | 2 | 299 |