Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015510.4 | NP_056325.2 | 3 | 323 | ||
XM_005256595.4 | XP_005256652.2 | 4 | 1117 | |||
XM_005256596.3 | XP_005256653.1 | 3 | 577 | |||
XM_011523786.2 | XP_011522088.1 | 4 | 1117 | |||
XM_017024425.1 | XP_016879914.1 | 3 | 435 | |||
XM_017024426.1 | XP_016879915.1 | 4 | 662 | |||
GenBank mRNA | AF151851.1 | - | 3 | 307 | ||
AL117567.1 | - | 3 | 310 | |||
BC004126.2 | - | 3 | 297 | 3937286 | ||
BC009679.2 | - | 3 | 296 | 3878039 | ||
BI460496.1 | - | 3 | 324 | |||
CR457124.1 | - | 3 | 279 |